2,4-Dichloro-L-phenylalanine 97%

Supplier: Ambeed
Warning

A506686-1G A506686-25G A506686-5G
103828-332EA 8 USD
103828-332 103839-434 77307-634 77307-636 77307-638
2,4-Dichloro-L-phenylalanine 97%
2,4-Dichloro-L-phenylalanine

Precaución: for Research Use Only

Formula: C₉H₉Cl₂NO₂
Boiling Pt: 368.1 °C (760 mm Hg)
Flash Pt: 176.4 °C
Storage Temperature: Ambient
MDL Number: MFCD01860882
CAS Number: 111119-36-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR