H-ORN(Z)-OH 97%

Supplier: Ambeed
Warning

Synonyms: Nδ-Benzyloxycarbonyl-L-ornithine

A226721-100G A226721-10G A226721-500G A226721-25G A226721-5G
103850-166EA 9 USD
103850-166 76722-134 76722-136 76722-138 77088-908 77088-910
H-ORN(Z)-OH 97%
H-ORN(Z)-OH

Precaución: for Research Use Only

Formula: C₁₃H₁₈N₂O₄
MW: 266.297 g/mol
Boiling Pt: 492.2 °C (760 mm Hg)
Flash Pt: 251.4 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00037220
CAS Number: 3304-51-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR