Homopiperazine 98%

Supplier: Ambeed
Danger

Synonyms: 1,4-Diazepane, 1,4-Diazacycloheptane, Perhydro-1,4-diazepine

A148165-10G A148165-1G A148165-25G A148165-5G
103803-882EA 7 USD
103803-882 103841-996 77095-740 77299-642 77317-428 77317-430
Homopiperazine 98%
Homopiperazine

Precaución: for Research Use Only

Formula: C₅H₁₂N₂
MW: 100.16 g/mol
Boiling Pt: 169 °C (760 mm Hg)
Melting Pt: 38…40 °C
Flash Pt: 61 °C
Storage Temperature: Ambient
MDL Number: MFCD00006933
CAS Number: 505-66-8
UN: 2923
ADR: 8,6.1,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR