(2S,3aS,7aS)-2-Carboxyoctahydroindole 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
(2S,3aS,7aS)-Octahydro-1H-indole-2-carboxylic acid, H-Oic-OH
A620435-100G
A620435-10G
A620435-1G
A620435-25G
103840-212EA
23
USD
103840-212
77254-382
77254-384
77307-964
77307-966
(2S,3aS,7aS)-2-Carboxyoctahydroindole 98%
(2S,3aS,7aS)-2-Carboxyoctahydroindole
Precaución: for Research Use Only
Formula:
C₉H₁₅NO₂ MW: 169.22 g/mol Flash Pt: 146.5 °C Storage Temperature: Ambient |
MDL Number:
MFCD07782125 CAS Number: 80875-98-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...