(2R,3S,5R)-5-Chloro-2-(((4-methylbenzoyl)oxy)methyl)tetrahydrofuran-3-yl 4-methylbenzoate 90%

Supplier: Ambeed
Warning

Synonyms: Hoffer's chlorosugar

A199575-1G A199575-250MG A199575-10G
103817-116EA 9 USD
103817-116 103847-740 77085-780 77274-768 77567-870
(2R,3S,5R)-5-Chloro-2-(((4-methylbenzoyl)oxy)methyl)tetrahydrofuran-3-yl 4-methylbenzoate 90%
(2R,3S,5R)-5-Chloro-2-(((4-methylbenzoyl)oxy)methyl)tetrahydrofuran-3-yl 4-methylbenzoate

Precaución: For research use only.

Formula: C₂₁H₂₁ClO₅
MW: 388.844 g/mol
Boiling Pt: 518.4 °C (760 mm Hg)
Flash Pt: 185.6 °C
Storage Temperature: Freezer
MDL Number: MFCD01630916
CAS Number: 4330-21-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR