L-Norvaline 97%

Supplier: Ambeed
Warning

Synonyms: (S)-(+)-Norvaline, L(+)-Norvaline, H-Nva-OH, (S)-2-Aminopentanoic acid

A190113-100G A190113-10G A190113-25G A190113-5G A190113-1G A190113-500G
103816-260EA 9 USD
103816-260 103846-856 76986-052 76986-054 77084-646 77084-648 77302-204 77683-493
L-Norvaline 97%
L-Norvaline

Precaución: For research use only.

Formula: C₅H₁₁NO₂
MW: 117.15 g/mol
Melting Pt: 191…196 °C
Flash Pt: 88.6 °C
Storage Temperature: Ambient
MDL Number: MFCD00064421
CAS Number: 6600-40-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR