N-Methyl-L-alanine 97%

Supplier: Ambeed
Warning

Synonyms: H-N-Me-Ala-OH, N-Me-Ala-OH

A285472-10G A285472-5G A285472-1G A285472-25G A285472-100G A285472-500G
103813-680EA 9 USD
103813-680 103834-814 76816-434 76837-352 77305-474 77305-476 77612-642 77672-472
N-Methyl-L-alanine 97%
N-Methyl-L-alanine

Precaución: for Research Use Only

Formula: C₄H₉NO₂
MW: 103.12 g/mol
Boiling Pt: 190.1 °C (760 mm Hg)
Flash Pt: 68.8 °C
Storage Temperature: Ambient
MDL Number: MFCD00037241
CAS Number: 3913-67-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR