H-Met-Otbu hydrochloride 97%

Supplier: Ambeed
Warning

Synonyms: H-Met-OtBu.HCl, H-Met-OtBu·HCl

A378499-1G A378499-250MG
103826-628EA 22 USD
103826-628 103837-662 76840-972 77115-474
H-Met-Otbu hydrochloride 97%
H-Met-Otbu hydrochloride

Precaución: for Research Use Only

Formula: C₉H₂₀ClNO₂S
MW: 241.774 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00038896
CAS Number: 91183-71-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR