L-Methionine methyl ester hydrochloride 98%

Supplier: Ambeed
Warning

A112696-100G A112696-10G A112696-25G A112696-5G
103823-918EA 7 USD
103823-918 77270-988 77270-990 77270-992 77310-728
L-Methionine methyl ester hydrochloride 98%
L-Methionine methyl ester hydrochloride

Precaución: for Research Use Only

Formula: C₆H₁₃NO₂S·HCl
Melting Pt: 151…153 °C
Flash Pt: 121.7 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00012491
CAS Number: 2491-18-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR