L-Methionine sulfoxide 97%

Supplier: Ambeed
Warning

Synonyms: H-Met(O)-OH, (2S)-2-Amino-4-(methylsulphinyl)butanoic acid

A108385-10G A108385-1G A108385-25G
103797-340EA 17 USD
103797-340 103823-120 76982-576 77283-718 77570-486
L-Methionine sulfoxide 97%
L-Methionine sulphoxide

Precaución: for Research Use Only

Formula: C₅H₁₁NO₃S
MW: 165.2 g/mol
Boiling Pt: 434.3 °C (760 mm Hg)
Melting Pt: 234…235 °C
Flash Pt: 216.4 °C
Storage Temperature: Ambient
MDL Number: MFCD00063093
CAS Number: 3226-65-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR