L-Lysine methyl ester dihydrochloride 98%

Supplier: Ambeed
Warning

Synonyms: H-Lys-OMe·2HCl, H-Lys-OMe.2HCl

A238818-100G A238818-10G A238818-25G A238818-5G A238818-500G
103851-044EA 7 USD
103851-044 77000-362 77000-364 77090-178 77090-180 77672-138
L-Lysine methyl ester dihydrochloride 98%
L-Lysine methyl ester dihydrochloride

Precaución: for Research Use Only

Formula: C₇H₁₆N₂O₂
MW: 233.13 g/mol
Boiling Pt: 243.9 °C (760 mm Hg)
Melting Pt: 213…215 °C
Storage Temperature: Freezer
MDL Number: MFCD00039067
CAS Number: 26348-70-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR