L(+)-Lysine dihydrochloride 96%
Supplier: Ambeed
|
Warning
|
A196617-25G
A196617-100G
103809-312EA
109
USD
103809-312
77085-438
77885-451
L(+)-Lysine dihydrochloride 96%
L(+)-Lysine dihydrochloride
Precaución: for Research Use Only
Formula:
C₆H₁₆Cl₂N₂O₂ Boiling Pt: 311.5 °C (760 mm Hg) Melting Pt: 200…206 °C Flash Pt: 142.2 °C Storage Temperature: Ambient |
MDL Number:
MFCD00066389 CAS Number: 657-26-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...