H-Lys(Z)-OtBu·HCl 97%

Supplier: Ambeed
Warning

Synonyms: H-Lys(Z)-OtBu·HCl, (S)-tert-Butyl 2-amino-6-(((benzyloxy)carbonyl)amino)hexanoate hydrochloride

A230959-10G A230959-25G A230959-5G A230959-1G A230959-100G A230959-500G
103810-738EA 6 USD
103810-738 103850-484 77031-036 77089-356 77089-358 77303-896 77600-846 77672-050
H-Lys(Z)-OtBu·HCl 97%
H-Lys(Z)-OtBu·HCl

Precaución: for Research Use Only

Formula: C₁₈H₂₉ClN₂O₄
Boiling Pt: 469.6 °C (760 mm Hg)
Flash Pt: 237.8 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00038898
CAS Number: 5978-22-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR