H-Lys(Fmoc)-OH 97%

Supplier: Ambeed
Warning

Synonyms: Nepsilon-Fmoc-L-lysine

A314461-1G A314461-10G A314461-25G A314461-100G
103825-036EA 10 USD
103825-036 103835-956 76838-772 76875-788 77306-114 77612-810
H-Lys(Fmoc)-OH 97%
H-Lys(Fmoc)-OH

Precaución: for Research Use Only

Formula: C₂₁H₂₄N₂O₄
MW: 368.433 g/mol
Boiling Pt: 609.9 °C (760 mm Hg)
Flash Pt: 322.6 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00038365
CAS Number: 84624-28-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR