H-Lys(boc)-OtBu·HCl 95%

Supplier: Ambeed
Warning

Synonyms: Nepsilon-Boc-L-lysine tert-butyl ester hydrochloride, H-Lys(Boc)-OtBu.HCl, H-Lys(Boc)-OtBu·HCl

A129957-10G A129957-250MG A129957-1G A129957-25G A129957-5G A129957-100G
103801-356EA 24 USD
103801-356 76996-316 76996-318 77313-768 77313-770 77313-772 77599-880
H-Lys(boc)-OtBu·HCl 95%
H-Lys(boc)-OtBu·HCl

Precaución: for Research Use Only

Formula: C₁₅H₃₁ClN₂O₄
MW: 338.873 g/mol
Flash Pt: 194.4 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00038899
CAS Number: 13288-57-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR