Nε-acetyl-L-lysine 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
Nε-Lys(Ac)-OH, (S)-6-Acetamido-2-aminohexanoic acid, L-Lys(Ac)-OH, H-Lys(Ac)-OH, Nepsilon-acetyl-L-lysine
A436907-250MG
A436907-25G
A436907-10G
103827-618EA
12
USD
103827-618
103838-684
76725-438
76842-422
77668-952
Nε-acetyl-L-lysine 97%
Nepsilon-acetyl-L-lysine
Precaución: for Research Use Only
Formula:
C₈H₁₆N₂O₃ MW: 188.23 g/mol Boiling Pt: 442 °C (760 mm Hg) Melting Pt: 250 °C Flash Pt: 221.1 °C Storage Temperature: Freezer |
MDL Number:
MFCD00002639 CAS Number: 692-04-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...