L-Leucine-tert-butyl ester hydrochloride 98%

Supplier: Ambeed
Warning

Synonyms: (S)-tert-Butyl 2-amino-4-methylpentanoate hydrochloride, H-Leu-OtBu·HCl, L-leucine 1,1-dimethylethyl ester hydrochloride, H-Leu-OtBu.HCl, TERT-BUTYL (2S)-2-AMINO-4-METHYLPENTANOATE HYDROCHLORIDE

A122711-100G A122711-10G A122711-25G A122711-500G
76797-460EA 98.67 USD
76797-460 76797-462 76797-464 77599-780
L-Leucine-tert-butyl ester hydrochloride 98%
L-Leucine-tert-butyl ester hydrochloride

Precaución: for Research Use Only

Formula: C₁₀H₂₁NO₂
MW: 223.74 g/mol
Flash Pt: 90.3 °C
Storage Temperature: Freezer
MDL Number: MFCD00038906
CAS Number: 2748-02-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR