L(+)-Leucinol 97%

Supplier: Ambeed
Warning

Synonyms: L-Leucinol, (S)-(+)-2-Amino-4-methylpentan-1-ol, (S)-2-Amino-4-methylpentan-1-ol

A586698-5G A586698-100G A586698-10G A586698-1G A586698-25G
103840-042EA 6 USD
103840-042 76844-184 77117-206 77117-208 77117-210 77307-884
L(+)-Leucinol 97%
L(+)-Leucinol

Precaución: for Research Use Only

Formula: C₆H₁₅NO
MW: 117.19 g/mol
Boiling Pt: 198…200 °C (760 mm Hg)
Flash Pt: 91 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00063676
CAS Number: 7533-40-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR