L-Isoleucinamide hydrochloride 97%

Supplier: Ambeed
Warning

Synonyms: H-Ile-NH2.HCl, H-Ile-NH₂·HCl

A147999-100G A147999-10G A147999-5G A147999-1G A147999-25G
103803-860EA 11 USD
103803-860 103841-976 77027-980 77027-982 77299-624 77317-404 77317-406
L-Isoleucinamide hydrochloride 97%
L-Isoleucinamide hydrochloride

Precaución: for Research Use Only

Formula: C₆H₁₄N₂O
MW: 166.65 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00058476
CAS Number: 10466-56-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR