(S)-(-)-Indoline-2-carboxylic acid 95%

Supplier: Ambeed
Warning

A375797-100G A375797-10G A375797-1G A375797-25G A375797-5G
103826-570EA 5 USD
103826-570 103837-594 76694-380 76694-382 76840-884 76840-886 76840-888
(S)-(-)-Indoline-2-carboxylic acid 95%
(S)-(-)-Indoline-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₉H₉NO₂
Melting Pt: 177 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00070578
CAS Number: 79815-20-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR