trans-4-Hydroxy-L(-)-proline 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
L-4-Hydroxyproline, (S)-(-)-trans-4-Hydroxyproline, L(-)-trans-4-Hydroxyproline, 4-Hydroxy-L-Proline, (2S,4R)-4-Hydroxypyrrolidine-2-carboxylic acid, H-Hyp-OH
A333728-25G
A333728-5G
A333728-100G
A333728-500G
A333728-1000G
103836-618EA
9
USD
103836-618
76839-624
76839-626
77167-450
77167-452
77683-995
trans-4-Hydroxy-L(-)-proline 97%
trans-4-Hydroxy-L(-)-proline
Precaución: For research use only.
Formula:
C₅H₉NO₃ MW: 131.13 g/mol Melting Pt: 273 °C Flash Pt: 168.6 °C Storage Temperature: Ambient |
MDL Number:
MFCD00064320 CAS Number: 51-35-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...