L-Homoserine 98%

Supplier: Ambeed
Warning

Synonyms: H-HomoSer-OH, H-HoSer-OH, H-Hser-OH, (S)-(-)-Homoserine, L(-)-Homoserine

A185112-25G A185112-5G A185112-100G A185112-100MG A185112-10G A185112-1G A185112-500G
103815-820EA 15 USD
103815-820 103846-386 77084-020 77084-022 77287-374 77287-376 77287-378 77301-948 77748-202
L-Homoserine 98%
L-Homoserine

Precaución: for Research Use Only

Formula: C₄H₉NO₃
MW: 119.12 g/mol
Boiling Pt: 368.7 °C (760 mm Hg)
Melting Pt: 203
Flash Pt: 176.8 °C
Storage Temperature: Ambient
MDL Number: MFCD00063090
CAS Number: 672-15-1
Merck Index: 13,04760

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR