L-Pipecolic acid 97%

Supplier: Ambeed
Warning

Synonyms: (S)-(-)-2-Piperidinecarboxylic Acid, H-HomoPro-OH, (S)-Piperidine-2-carboxylic acid, L-Pipecolinic Acid, L-Homoproline

A127260-100G A127260-10G A127260-1G A127260-25G A127260-5G
103800-940EA 5 USD
103800-940 77161-650 77161-652 77298-196 77298-198 77298-200
L-Pipecolic acid 97%
L-Pipecolic acid

Precaución: for Research Use Only

Formula: C₆H₁₁NO₂
MW: 129.16 g/mol
Boiling Pt: 265.8 °C (760 mm Hg)
Flash Pt: 114.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00005981
CAS Number: 3105-95-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR