(S)-2-Amino-4-phenylbutanoic acid 98%

Supplier: Ambeed
Warning

Synonyms: H-HoPhe-OH, H-Homophe-OH, (S)-2-Amino-4-phenylbutyric Acid

A100606-100G A100606-1G A100606-5G A100606-25G A100606-500G
103821-224EA 7 USD
103821-224 76687-004 76687-006 77296-776 77548-890 77623-816
(S)-2-Amino-4-phenylbutanoic acid 98%
(S)-2-Amino-4-phenylbutanoic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₃NO₂
MW: 179.22 g/mol
Boiling Pt: 324.8 °C (760 mm Hg)
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00002619
CAS Number: 943-73-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR