Glycyl-L-Proline

Supplier: Ambeed

A134704-250MG A134704-5G A134704-1G A134704-100MG
103802-042EA 49 USD
103802-042 103854-890 77109-452 77298-724 77314-814 77611-204
Glycyl-L-Proline
Proteins and Peptides

  • Boiling pointL 11.3 °C (760 mm Hg)
  • Flash point: 202.6 °C
  • GHS symbol and signal word: GHS07 and warning


Precaución: Keep in a dark place, inert atmosphere, store in freezer, under –20 °C.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR