2-Amino-N-methylacetamide hydrochloride 97%
Supplier: Ambeed
|
Warning
|
A117128-100G
A117128-10G
A117128-250MG
A117128-25G
A117128-1G
A117128-5G
103798-834EA
9
USD
103798-834
103824-710
76797-122
76797-124
76797-126
76797-128
77311-494
77311-496
2-Amino-N-methylacetamide hydrochloride 97%
2-Amino-N-methylacetamide hydrochloride
Precaución: for Research Use Only
Formula:
C₃H₉ClN₂O Melting Pt: 153.5…156 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00153440 CAS Number: 49755-94-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...