Glycinamide hydrochloride (H-Gly-NH2.HCl) 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
H-Gly-NH2.HCl
A206377-1000G
A206377-100G
A206377-10G
A206377-500G
A206377-25G
103848-378EA
7
USD
103848-378
76999-508
76999-510
76999-512
76999-514
77086-628
Glycinamide hydrochloride (H-Gly-NH2.HCl) 98%
Glycinamide hydrochloride (H-Gly-NH2.HCl)
Precaución: for Research Use Only
Formula:
C₂H₇ClN₂O MW: 110.54 g/mol Boiling Pt: 281.3 °C (760 mm Hg) Melting Pt: 204 °C Storage Temperature: Ambient |
MDL Number:
MFCD00013008 CAS Number: 1668-10-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...