H-Glu-otbu 95% (contains < 9% H₂O)

Supplier: Ambeed
Warning

Synonyms: (S)-4-Amino-5-(tert-butoxy)-5-oxopentanoic acid, L-Glutamic acid 1-tert-butyl ester, 1-tert-Butyl L-Glutamate, H-Glu-OtBu, L-Glutamic acid 1-tert-Butyl ester

A198934-10G A198934-1G A198934-5G
103817-058EA 6 USD
103817-058 103847-684 77097-374 77302-604 77302-606
H-Glu-otbu 95% (contains < 9% H₂O)
H-Glu-otbu

Precaución: for Research Use Only

Formula: C₉H₁₇NO₄
MW: 203.238 g/mol
Melting Pt: 140 °C
Flash Pt: 151.5 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00038562
CAS Number: 45120-30-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR