1-Benzyl-L-glutamate 97%

Supplier: Ambeed
Warning

Synonyms: L-Glutamic Acid 1-Benzyl Ester, H-Glu-OBzl, (S)-4-Amino-5-(benzyloxy)-5-oxopentanoic acid

A265153-1G A265153-25G A265153-5G A265153-100G
103812-710EA 8 USD
103812-710 103852-604 77092-272 77092-274 77304-912 77668-138
1-Benzyl-L-glutamate 97%
1-Benzyl-L-glutamate

Precaución: for Research Use Only

Formula: C₁₂H₁₅NO₄
Boiling Pt: 416 °C (760 mm Hg)
Melting Pt: 157 °C
Storage Temperature: Freezer
MDL Number: MFCD00037312
CAS Number: 13030-09-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR