H-Glu(OtBu)-OtBu·HCl 98%

Supplier: Ambeed
Warning

Synonyms: H-Glu(OtBu)-OtBu·HCl, L-Glutamic acid di-tert-butyl ester hydrochloride, H-Glu(OtBu)-OtBu.HCl

A386701-10G A386701-1G A386701-25G A386701-5G A386701-500G A386701-100G
103826-806EA 5 USD
103826-806 103837-848 76724-940 76841-204 76841-206 76841-208 77568-790 77601-702
H-Glu(OtBu)-OtBu·HCl 98%
H-Glu(OtBu)-OtBu·HCl

Precaución: For research use only.

Formula: C₁₃H₂₆ClNO₄
MW: 295.804 g/mol
Flash Pt: 78.1 °C
Storage Temperature: Freezer
MDL Number: MFCD00058003
CAS Number: 32677-01-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR