5-tert-Butyl-1-methyl-L-glutamate hydrochloride 98%

Supplier: Ambeed
Warning

Synonyms: L-Glutamic Acid 5-tert-Butyl 1-Methyl Ester Hydrochloride, H-Glu(OtBu)-OMe·HCl, H-Glu(OtBu)-OMe.HCl

A147316-1G A147316-5G A147316-10G A147316-100G
103803-760EA 9 USD
103803-760 103841-860 77317-230 77317-232 77567-034 77624-256
5-tert-Butyl-1-methyl-L-glutamate hydrochloride 98%
5-tert-Butyl-1-methyl-L-glutamate hydrochloride

Precaución: For research use only.

Formula: C₁₀H₁₉NO₄
MW: 253.72 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00153438
CAS Number: 6234-01-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR