Dimethyl-L-glutamate hydrochloride 98%

Supplier: Ambeed
Warning

Synonyms: L-Glutamic Acid Dimethyl Ester Hydrochloride, H-Glu(OMe)-OMe.HCl

A187261-100G A187261-10G A187261-500G A187261-25G
103809-212EA 283 USD
103809-212 77111-622 77111-624 77111-626 77302-068
Dimethyl-L-glutamate hydrochloride 98%
Dimethyl-L-glutamate hydrochloride

Precaución: for Research Use Only

Formula: C₇H₁₃NO₄
Melting Pt: 99…103 °C
Flash Pt: 76.7 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00038879
CAS Number: 23150-65-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR