L-Glutamic acid 5-benzyl ester 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
γ-Benzyl-L-Glutamate, 5-Benzyl L-Glutamate, gamma-Benzyl L-glutamate, H-Glu(OBzl)-OH
A182358-100G
A182358-10G
A182358-5G
103846-072EA
6
USD
103846-072
77274-118
77274-120
77301-778
L-Glutamic acid 5-benzyl ester 97%
L-Glutamic acid 5-benzyl ester
Precaución: for Research Use Only
Formula:
C₁₂H₁₅NO₄ Boiling Pt: 426.1 °C (760 mm Hg) Melting Pt: 181…182 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002633 CAS Number: 1676-73-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...