H-Glu(OBzl)-OBzl.TosOH 98%

Supplier: Ambeed
Warning

Synonyms: L-Glutamic acid dibenzyl ester p-toluenesulphonate, H-Glu(OBzl)-OBzl·TosOH, H-Glu(OBzl)-OBzl·p-tosylate

A108723-100G A108723-10G A108723-25G A108723-5G
103808-004EA 350 USD
103808-004 103823-168 76995-082 76995-084 76995-086 77310-000
H-Glu(OBzl)-OBzl.TosOH 98%
H-Glu(OBzl)-OBzl.TosOH

Precaución: for Research Use Only

Formula: C₂₆H₂₉NO₇S
MW: 499.578 g/mol
Boiling Pt: 453.5 °C (760 mm Hg)
Flash Pt: 165.6 °C
Storage Temperature: Ambient
MDL Number: MFCD00038980
CAS Number: 2791-84-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR