N'-Trityl-L-glutamine 97%

Supplier: Ambeed
Warning

Synonyms: H-Gln(Trt)-OH

A100087-25G A100087-1G
103795-238EA 9 USD
103795-238 103820-958 77025-100 77296-562
N'-Trityl-L-glutamine 97%
N'-Trityl-L-glutamine

Precaución: for Research Use Only

Formula: C₂₄H₂₄N₂O₃
MW: 388.467 g/mol
Boiling Pt: 658 °C (760 mm Hg)
Flash Pt: 351.7 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00237060
CAS Number: 102747-84-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR