1-Hexylboronic acid 96%

Supplier: Ambeed
Warning

A413956-1G A413956-25G A413956-10G A413956-5G
103827-268EA 14 USD
103827-268 103838-312 76841-876 76841-878 77155-764 77307-168
1-Hexylboronic acid 96%
1-Hexylboronic acid

Precaución: for Research Use Only

Formula: C₆H₁₅BO₂
Boiling Pt: 226.5 °C (760 mm Hg)
Melting Pt: 85…90 °C
Storage Temperature: Freezer
MDL Number: MFCD01074641
CAS Number: 16343-08-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR