Hexyl 2-(4-(diethylamino)-2-hydroxybenzoyl)benzoate 98%

Supplier: Ambeed
Warning

Synonyms: Diethylamino hydroxybenzoyl hexyl benzoate

A107883-10G A107883-25G A107883-500G A107883-5G
103823-026EA 12 USD
103823-026 76821-644 76821-646 76821-648 77283-660
Hexyl 2-(4-(diethylamino)-2-hydroxybenzoyl)benzoate 98%
Hexyl 2-(4-(diethylamino)-2-hydroxybenzoyl)benzoate

Precaución: for Research Use Only

Formula: C₂₄H₃₁NO₄
MW: 397.51 g/mol
Boiling Pt: 524.8 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD11616752
CAS Number: 302776-68-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR