H-D-Val-OtBu·HCl 97%

Supplier: Ambeed
Warning

Synonyms: H-D-Val-OtBu.HCl, D-Valine tert-butyl ester hydrochloride, H-D-Val-OtBu·HCl

A225227-10G A225227-25G A225227-5G
103850-058EA 9 USD
103850-058 77000-038 77088-740 77088-742
H-D-Val-OtBu·HCl 97%
H-D-Val-OtBu·HCl

Precaución: for Research Use Only

Formula: C₉H₂₀ClNO₂
MW: 209.714 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00237308
CAS Number: 104944-18-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR