D-(+)-Tryptophan 97%
Supplier: Ambeed
|
Warning
|
A101619-100G
A101619-10G
A101619-25G
A101619-500G
A101619-5G
A101619-1000G
103807-806EA
207
USD
103807-806
103821-742
77270-252
77270-254
77270-256
77270-258
77282-772
77297-168
D-(+)-Tryptophan 97%
D-(+)-Tryptophan
Precaución: for Research Use Only
Formula:
C₁₁H₁₂N₂O₂ Boiling Pt: 447.9 °C (760 mm Hg) Melting Pt: 282…285 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005647 CAS Number: 153-94-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...