D-Tryptophan benzyl ester hydrochloride 97%

Supplier: Ambeed
Warning

Synonyms: H-D-Trp-OBzl·HCl, H-D-Trp-OBzl.HCl

A452458-10G A452458-1G A452458-25G A452458-5G
103827-798EA 9 USD
103827-798 103838-868 76804-526 77307-360 77307-362 77307-364
D-Tryptophan benzyl ester hydrochloride 97%
D-Tryptophan benzyl ester hydrochloride

Precaución: for Research Use Only

Formula: C₁₈H₁₈N₂O₂
MW: 330.81 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00237293
CAS Number: 22839-16-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR