D-tert-Leucine (H-D-Tle-OH) 97%

Supplier: Ambeed
Warning

A238633-25G A238633-100G A238633-10G A238633-500G A238633-250MG
103811-242EA 6 USD
103811-242 103851-028 77090-152 77153-040 77153-042 77153-044 77892-417
D-tert-Leucine (H-D-Tle-OH) 97%
D-tert-Leucine (H-D-Tle-OH)

Precaución: for Research Use Only

Formula: C₆H₁₃NO₂
Melting Pt: ≥300 °C
Storage Temperature: Ambient
MDL Number: MFCD00004265
CAS Number: 26782-71-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR