D-Threonine 98%
Supplier: Ambeed
|
Warning
|
A155576-100G
A155576-10G
A155576-25G
A155576-5G
103842-978EA
6
USD
103842-978
77110-476
77110-478
77300-132
77300-134
D-Threonine 98%
D-Threonine
Precaución: for Research Use Only
Formula:
C₄H₉NO₃ Boiling Pt: 345.8 °C (760 mm Hg) Melting Pt: 274 °C Flash Pt: 162.9 °C Storage Temperature: Ambient |
MDL Number:
MFCD00064269 CAS Number: 632-20-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...