H-D-Thr(tBu)-OH 97%

Supplier: Ambeed
Warning

A304309-10G A304309-5G
103835-580EA 15 USD
103835-580 77194-498 77305-900
H-D-Thr(tBu)-OH 97%
H-D-Thr(tBu)-OH

Precaución: for Research Use Only

Formula: C₈H₁₇NO₃
Storage Temperature: Refrigerator
MDL Number: MFCD00171400
CAS Number: 201274-81-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR