D-(-)-Serine 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
(R)-(-)-Serine, H-D-Ser-OH
A111308-100G
A111308-10G
A111308-1G
A111308-25G
A111308-500G
A111308-5G
A111308-1000G
103823-628EA
6
USD
103823-628
76970-798
76970-800
76970-802
76970-804
76970-806
77284-104
77670-578
D-(-)-Serine 95%
D-(-)-Serine
Precaución: for Research Use Only
Formula:
C₃H₇NO₃ MW: 105.09 g/mol Boiling Pt: 394.8 °C (760 mm Hg) Melting Pt: 235 Flash Pt: 192.6 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004269 CAS Number: 312-84-5 Merck Index: 12,08604 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...