H-D-Ser(tBu)-OtBu·HCl 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
H-D-Ser(tBu)-OtBu.HCl
A103033-10G
A103033-250MG
A103033-1G
A103033-5G
103796-470EA
6
USD
103796-470
103822-172
76796-334
76796-336
77309-040
77309-042
H-D-Ser(tBu)-OtBu·HCl 97%
H-D-Ser(tBu)-OtBu·HCl
Precaución: for Research Use Only
Formula:
C₁₁H₂₃NO₃·HCl MW: 253.8 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD00153462 CAS Number: 179559-35-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...