D-(+)-Proline 98%

Supplier: Ambeed
Warning

Synonyms: H-D-Pro-OH, (R)-(+)-Proline, D-Proline

A121722-100G A121722-10G A121722-25G A121722-500G
103808-240EA 167 USD
103808-240 77271-540 77271-542 77271-544 77271-546
D-(+)-Proline 98%
D-(+)-Proline

Precaución: for Research Use Only

Formula: C₅H₉NO₂
MW: 115.13 g/mol
Boiling Pt: 252.2 °C (760 mm Hg)
Melting Pt: 223 °C
Flash Pt: 106.3 °C
Storage Temperature: Ambient
MDL Number: MFCD00064317
CAS Number: 344-25-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR