D-(-)-α-(4-Hydroxyphenyl)glycine 98%

Supplier: Ambeed
Warning

Synonyms: D(-)-4-Hydroxyphenylglycine, H-D-Phg(4-OH)-OH, (R)-(-)-2-(4-Hydroxyphenyl)glycine, (-)-4-Hydroxy-D-phenylglycine, D-(-)-p-Hydroxyphenylglycine

A151368-100G A151368-10G A151368-500G A151368-1000G A151368-25G
103808-754EA 93 USD
103808-754 77247-280 77247-282 77247-284 77317-958 77317-960
D-(-)-α-(4-Hydroxyphenyl)glycine 98%
D(-)-α-(4-Hydroxyphenyl)glycine

Precaución: for Research Use Only

Formula: C₈H₉NO₃
MW: 167.16 g/mol
Boiling Pt: 365.8 °C (760 mm Hg)
Melting Pt: 240 °C
Flash Pt: 223.7 °C
Storage Temperature: Ambient
MDL Number: MFCD00004262
CAS Number: 22818-40-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR