(±)-3-Amino-3-phenylpropionic acid 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
DL-β-Phenylalanine, 3-Amino-3-phenylpropionic acid, H-DL-β-Phe-OH, β-Aminohydrocinnamic acid
A189586-1G
A189586-10G
A189586-100G
A189586-500G
A189586-1000G
103816-204EA
5
USD
103816-204
103846-798
77084-570
77248-528
77615-602
77615-604
77746-216
(±)-3-Amino-3-phenylpropionic acid 97%
(±)-3-Amino-3-phenylpropionic acid
Precaución: for Research Use Only
Formula:
C₉H₁₁NO₂ MW: 165.19 g/mol Boiling Pt: 307.5 °C (760 mm Hg) Melting Pt: 222 °C Flash Pt: 139.8 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008064 CAS Number: 614-19-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...