H-D-Lys(Z)-OH 97%

Supplier: Ambeed
Warning

Synonyms: H-D-Lys(Cbz)-OH, Nepsilon-Benzyloxycarbonyl-D-lysine, Nepsilon-Cbz-D-lysine, H-D-Lys(Z)-OH, Nepsilon-Carbobenzoxy-D-lysine

A328111-1G A328111-5G A328111-10G
103825-494EA 14 USD
103825-494 103836-448 76839-408 76839-410 77099-962
H-D-Lys(Z)-OH 97%
H-D-Lys(Z)-OH

Precaución: for Research Use Only

Formula: C₁₄H₂₀N₂O₄
MW: 280.324 g/mol
Melting Pt: 229 °C
Flash Pt: 255.9 °C
Storage Temperature: Ambient
MDL Number: MFCD00063120
CAS Number: 34404-32-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR