H-D-Lys(Boc)-OMe·HCl 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
H-D-Lys(Boc)-OMe.HCl
A335163-1G
A335163-5G
A335163-10G
A335163-250MG
103825-686EA
90
USD
103825-686
103836-656
76839-668
76839-670
76989-334
76989-336
H-D-Lys(Boc)-OMe·HCl 95%
H-D-Lys(Boc)-OMe·HCl
Precaución: for Research Use Only
Formula:
C₁₂H₂₄N₂O₄·HCl MW: 296.8 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD00180720 CAS Number: 66494-53-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...